Order Entry
Puerto Rico
ContactUsLinkComponent
MA EPH Aromatic Hydrocarbon Standard (17 components), Restek
MA EPH Aromatic Hydrocarbon Standard (17 components), Restek
Catalog # RK31458
Supplier:  Restek
CAS Number:  
undefined
MA EPH Aromatic Hydrocarbon Standard (17 components), Restek
Catalog # RK31458
Supplier:  Restek
Supplier Number:  31458
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml each in methylene chloride
  • Description:
    MA EPH Aromatic Hydrocarbon Standard
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK31458
  • Certification:
    CRM
  • Supplier no.:
    31458

Specifications

About this item

Contains: Acenaphthene, acenaphthylene, anthracene, benz(a)anthracene, benzo(a)pyrene, benzo(b)fluoranthene, benzo(k)fluoranthene, benzo(ghi)perylene, chrysene, dibenz(a,h)anthracene, fluoranthene, fluorene, indeno(1,2,3-cd)pyrene, 2-methylnaphthalene, naphthalene, phenanthrene, pyrene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.