610 PAH Calibration Mix A (16 components), Restek
RK31264
: Restek
:
- Concentration:In methylene chloride
- Description:610 PAH Calibration Mix A
- Size:1 ml/Ampule
- Cat. No.:RK31264
- Certification:CRM
- Supplier no.:31264
610 PAH calibration mix A contains acenaphthene (1000 μg/ml), acenaphthylene (1000 μg/ml),anthracene (1000 μg/ml), benz(a)anthracene (500 μg/ml), benzo(a)pyrene (500 μg/ml), benzo(b)fluoranthene (500 μg/ml)
benzo(k)fluoranthene (500 μg/ml), benzo(ghi)perylene (500 μg/ml), chrysene (500 μg/ml), dibenz(a,h)anthracene (500 μg/ml), fluoranthene (500 μg/ml), fluorene (1000 μg/ml), indeno(1,2,3-cd)pyrene (500 μg/ml), naphthalene (1000 μg/ml), phenanthrene (500 μg/ml), pyrene ( 500 μg/ml).
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.