Order Entry
Puerto Rico
ContactUsLinkComponent
SV Calibration Mix #2 (15 Components), Restek
SV Calibration Mix #2 (15 Components), Restek
Catalog # RK31008
Supplier:  Restek
CAS Number:  
undefined
SV Calibration Mix #2 (15 Components), Restek
Catalog # RK31008
Supplier:  Restek
Supplier Number:  31008
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Description:
    SV Calibration Mix #2
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK31008
  • Certification:
    CRM
  • Supplier no.:
    31008

Specifications

About this item

Contains: Benzoic acid (65-85-0); 4-Chloro-3-methylphenol (59-50-7); 2-Chlorophenol (95-57-8); 2,4-Dichlorophenol (120-83-2); 2,4-Dimethylphenol (105-67-9); 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol) (534-52-1); 2,4-Dinitrophenol (51-28-5); 2-Methylphenol (o-cresol) (95-48-7); 4-Methylphenol (p-cresol) (106-44-5); 2-Nitrophenol (88-75-5); 4-Nitrophenol (100-02-7); Pentachlorophenol (87-86-5); Phenol (108-95-2); 2,4,5-Trichlorophenol (95-95-4); 2,4,6-Trichlorophenol (88-06-2).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.