WA VPH Marker Standard (9 Components), Restek
Catalog # RK30450
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in P and T methanol
- Description:WA VPH Marker Standard
- Size:1 ml/Ampule
- Cat. No.:RK30450
- Certification:CRM
- Supplier no.:30450
Specifications
About this item
Contains: (C5) n-Pentane (109-66-0); (C6) n-Hexane (110-54-3); (C8) n-Octane (111-65-9); (C10) n-Decane (124-18-5); (C12) n-Dodecane (112-40-3); 1-Methylnaphthalene (90-12-0); Naphthalene (91-20-3); Toluene (108-88-3); 1,2,3-Trimethylbenzene (526-73-8).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.