1,2,4-Trimethylbenzene, Restek
RK30422
: Restek
:
- Concentration:2000 µg/ml in P and T methanol
- CAS No.:95-63-6
- Description:1,2,4-Trimethylbenzene
- Size:1 ml/Ampule
- Formula:C₉H₁₂
- Description:1,2,4-Trimethylbenzene
- Cat. No.:RK30422
- Certification:CRM
- Supplier no.:30422
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.