Methyl Methacrylate, Restek
Catalog # RK30299
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in P and T methanol
- CAS No.:80-62-6
- Description:Methyl methacrylate
- Size:1 ml/Ampule
- Formula:C₅H₈O₂
- Description:Methyl methacrylate
- Cat. No.:RK30299
- Certification:CRM
- Supplier no.:30299
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.