Acrylonitrile, Restek
Catalog # RK30246
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in P and T methanol
- CAS No.:107-13-1
- Description:Acrylonitrile
- Size:1 ml/Ampule
- Formula:C₃H₃N
- Description:Acrylonitrile
- Cat. No.:RK30246
- Certification:CRM
- Supplier no.:30246
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.