8240 Nitriles Mix (7 components), Restek
Catalog # RK30215
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in P and T methanol
- Description:8240 Nitriles Mix
- Size:1 ml/Ampule
- Cat. No.:RK30215
- Certification:CRM
- Supplier no.:30215
Specifications
About this item
Mix contains: Acrylonitrile (107-13-1); Ethyl methacrylate (97-63-2); Malononitrile (109-77-3); Methacrylonitrile (126-98-7); Methyl methacrylate (80-62-6); Propionitrile (107-12-0); Styrene (100-42-5).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.