Order Entry
Puerto Rico
ContactUsLinkComponent
8240 Alcohols Mix (5 components), Restek
8240 Alcohols Mix (5 components), Restek
Catalog # RK30214
Supplier:  Restek
CAS Number:  
undefined
8240 Alcohols Mix (5 components), Restek
Catalog # RK30214
Supplier:  Restek
Supplier Number:  30214
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in P and T methanol
  • Description:
    8240 Alcohols Mix
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK30214
  • Certification:
    CRM
  • Supplier no.:
    30214

Specifications

About this item

Mix contains: Allyl alcohol (2-propen-1-ol) (107-18-6); 2-Chloroethanol (107-07-3); Ethanol (64-17-5); Isobutyl alcohol (78-83-1); Propargyl alcohol (107-19-7).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.