ContactUsLinkComponent
L/C VOA Internal Standard Mix (3 Components), Restek
  RK30091
 :  Restek
 :  
undefined
L/C VOA Internal Standard Mix (3 Components), Restek
  RK30091
 :  Restek
 :  30091
 :  
  • Concentration:
    2500 µg/ml each in P and T methanol
  • Description:
    L/C VOA Internal Standard Mix
  • Size:
    1 ml/Ampule
  • Cat. No.:
    RK30091
  • Certification:
    CRM
  • Supplier no.:
    30091
Mix contains: Chlorobenzene-d5 (3114-55-4), 1,4-Dichlorobenzene-d4 (3855-82-1), 1,4-Difluorobenzene (540-36-3).
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.