Order Entry
Puerto Rico
ContactUsLinkComponent
FID Performance Evaluation Standard (3 components), Restek
FID Performance Evaluation Standard (3 components), Restek
Catalog # 10058-060
Supplier:  Restek
CAS Number:  
undefined
FID Performance Evaluation Standard (3 components), Restek
Catalog # 10058-060
Supplier:  Restek
Supplier Number:  33908
CAS Number:  

Specifications

  • Concentration:
    0.03% w/w each in hexane
  • Description:
    FID Performance Evaluation Standard
  • Size:
    1 ml/Ampule
  • Cat. No.:
    10058-060
  • Certification:
    CRM
  • Supplier no.:
    33908

Specifications

About this item

Contains: (C14) n-Tetradecane (629-59-4); (C15) n-Pentadecane (629-62-9); (C16) n-Hexadecane (544-76-3)