Order Entry
Puerto Rico
ContactUsLinkComponent
 
LC OQ Linearity Test Mix Kit, Restek
  10058-056
 :  Restek
 :  
undefined
LC OQ Linearity Test Mix Kit, Restek
  10058-056
 :  Restek
 :  31805
 :  

 

  • Concentration:
    1 ml each of these mixtures
  • Description:
    LC OQ Linearity Test Mix Kit
  • Size:
    Kit
  • Cat. No.:
    10058-056
  • Certification:
    CRM
  • Supplier no.:
    31805

 

 

Contains:

  • Caffeine at 5, 25, 125, 250, 500 µg/mL in water in a five-ampul kit.
  • Simple check of a detector’s ability to produce a linear response to varied concentrations.
  • Suitable for clear indication of linear range.