Order Entry
Puerto Rico
ContactUsLinkComponent
EZ-Yeast™ Transformation Kit, MP Biomedicals
EZ-Yeast™ Transformation Kit, MP Biomedicals
Catalog # IC112100200
Supplier:  MP Biomedicals
CAS Number:  
undefined
EZ-Yeast™ Transformation Kit, MP Biomedicals
Catalog # IC112100200
Supplier:  MP Biomedicals
Supplier Number:  112100200
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Transformation Kits, with EZ-Transformation Solution, Carrier DNA
  • Storage Conditions:
    Store at Room Temperature
  • Application:
    Cloning and Expression, Transformation, Yeast Cell Culture, Cell Viability Analysis, Microbiological Analysis
  • Cat. No.:
    IC112100200
  • Supplier no.:
    112100200

Specifications

About this item

The EZ-Yeast™ Transformation Kit is designed for high-throughput transformation with efficiencies up to 103 transformants per µg plasmid DNA.

  • Ideal where large number of transformations must be performed
  • Perfect for a quick analysis of potentially positive two-hybrid colonies
  • Does not require fully competent cells prior to transormation

The EZ-Yeast Transformation Kit is designed for high throughput transformations

Kits are for Cloning and Expression, Transformation, Yeast Cell Culture, Cell Viability Analysis, Microbiological Analysis.

It is ideal for cases where a large number of transformations must be performed, but only a few transformants are needed. Simply remove a 3-4 mm colony or 0.5 mL overnight culture to a new tube, add vector and carrier DNA, incubate for 30 minutes and plate on selective media.