Order Entry
Puerto Rico
ContactUsLinkComponent
 
Benzene, Restek
  75835-382
 :  Restek
 :  
Benzene, Restek
  75835-382
 :  Restek
 :  35262
 :  

 

  • Concentration:
    250 µg/ml in methylene chloride
  • CAS No.:
    71-43-2
  • Description:
    Benzene
  • Size:
    1 ml/Ampule
  • Formula:
    C₆H₆
  • Description:
    Benzene
  • Cat. No.:
    75835-382
  • Certification:
    CRM
  • Supplier no.:
    35262

 

 

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.