Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
CoolCaddy™ Cold WorkStation

CoolCaddy™ Cold WorkStation

CoolCaddy™ cold workstation, 21×17×4.5 cm
Catalog # 470228-446
Supplier:   MTC BIO, INC.
Supplier #:  R4015
CoolCaddy™ Cold WorkStation
Catalog # 470228-446
Supplier:  MTC BIO, INC.
CoolCaddy™ Cold WorkStation
CoolCaddy™ Cold WorkStation

CoolCaddy™ cold workstation, 21×17×4.5 cm

Price
$183.00
Log in for availability
Each
Product Details & Documents

Keeps your samples cool while you experiment.

  • Maintains 1 to 4 °C for up to 5 hours
  • Keeps samples cool but not frozen
  • Eliminates the need for crushed ice
  • Accomodates most small tube types

Keeps your reagents and samples cool on the lab bench without the use of crushed ice.

Simply freeze overnight and its ready to hold 1 to 4 °C for up to 5 hours, keeping your Microtubes, Cryovials, PCR Tubes/Plate and Reagent Reservoirs cold (but not frozen) while you work.

Specifications
  • Description:
    CoolCaddy™ cold workstation
  • Dimensions:
    21×17×4.5 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.