Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce ProFound™ GST Protein Interaction Pull Down Kit, Thermo Scientific
Pierce ProFound™ GST Protein Interaction Pull Down Kit, Thermo Scientific
Catalog # PI21516
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce ProFound™ GST Protein Interaction Pull Down Kit, Thermo Scientific
Catalog # PI21516
Supplier:  Invitrogen
Supplier Number:  21516
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    GST-Protein:Protein Interaction Kit
  • Cat. No.:
    PI21516
  • No. of reactions:
    25 Pull-down reactions, each using 25 µl of resin
  • Supplier no.:
    21516

Specifications

About this item

The Pierce™ GST Interaction Pull-Down Kit contains the necessary components to capture and purify proteins that interact with GST-tagged fusion proteins.

  • Complete kits provide all components and detailed protocols for purifying protein:protein interactions
  • No special equipment needed – use common laboratory equipment and reagents
  • Microcentrifuge spin columns facilitate simple and efficient manipulation of agarose beads, including simple processing of multiple samples
  • Instructions include protocols for use with bait and prey proteins expressed from a variety of sample types
  • Cobalt chelate resin is more specific for histidine-tagged fusion proteins than nickel resins, resulting in less non-specific binding
  • Binds 10 to 25 mg of histidine-tagged fusion protein per mL of resin

Applications include discovering a new protein:protein interaction from a cell lysate; confirming a putative interaction from a cell lysate or with a previously purified protein; extracting protein:protein interaction information from in vitro transcription/translation lysates.

Delivery: Complete kit includes Cobalt chelate agarose resin, (750 µL); Pull-Down Lysis Buffer, (250 mL); Lysis buffer, (250 mL); TBS pack (makes 500 mL); Spin columns and accessories, (27 columns); and Microcentrifuge collection tubes, (100 tubes).