Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Buffer Solution, Sodium Phosphate, pH 7.4, Moltox

Buffer Solution, Sodium Phosphate, pH 7.4, Moltox

Marketsource
Buffer Solution, Sodium Phosphate, pH 7.4, Moltox
Marketsource
Certificates
Buffer Solution, Sodium Phosphate, pH 7.4, Moltox

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Product Family Options

Product Information

  • Size
    Pack type
    pH
    pH buffer type
    Availability
    Price
  • 100190-550
    Sodium Phosphate Buffer 0.1 M
    Sodium Phosphate Buffer 0.1 M
    100 ml
    Bottle
    7.4
    Phosphate
    Log in for availability
    Log in for availability
    Each
    Each - $42.31
    $42.31
    Each
       $42.31

    Supplier Number: 26-543.039
    Specifications:
    Marketsource
    Marketsource
  • 100190-634
    Sodium Phosphate Buffer 2.0 M
    Sodium Phosphate Buffer 2.0 M
    500 ml
    Bottle
    7.4
    Phosphate
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $132.35
    $132.35
    Each (500ml)
       $132.35

    Supplier Number: 26-516.5
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.