Order Entry
United States
ContactUsLinkComponent
Chlorine Calibration Gas, Draeger Inc
Chlorine Calibration Gas, Draeger Inc
Catalog # 34330-696
Supplier:  Draeger
CAS Number:  
Chlorine Calibration Gas, Draeger Inc
Catalog # 34330-696
Supplier:  Draeger
Supplier Number:  4594964
CAS Number:  

Specifications

  • Description:
    Chlorine Calibration Gas for Nitrogen Gas Detection, 5 ppm, 500 PSI
  • Environmentally Preferable:
  • Cat. No.:
    34330-696
  • Supplier no.:
    4594964

Specifications

About this item