Order Entry
Puerto Rico
ContactUsLinkComponent
ID-Vit® Biotin Microtiter Plate Assay, Eagle Biosciences
ID-Vit® Biotin Microtiter Plate Assay, Eagle Biosciences
Catalog # 102993-722
Supplier:  Eagle Biosciences
CAS Number:  
ID-Vit® Biotin Microtiter Plate Assay, Eagle Biosciences
Catalog # 102993-722
Supplier:  Eagle Biosciences
Supplier Number:  KIFR007
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    50 µl
  • Cat. No.:
    102993-722
  • Sensitivity:
    0.02 µg/l
  • No. of wells:
    1x96 wells
  • Storage temperature:
    +2…+8 °C
  • Sample type:
    Serum
  • Application:
    ELISA
  • Range:
    0.02 - 0.18 µg/l
  • Supplier no.:
    KIF007

Specifications

About this item

Biotin Assay kit is a microtiter plate test kit based on a microbiological assay which measures the total free biotin content in serum.

  • Incubation Time: 48 hours

The test kit contains all required reagents, e.g. standard, medium and microtiter plate coated with a specific microorganism, sufficient for 96 determinations including standard curves. An ELISA reader is required for evaluation of the biotin con­tent. It can be used in human and veterinary medicine and in research only.

Delivery: It includes a Lactobacillus plantarum - precoated microtiter plate (ready to use
), Water 30 mL, Biotin assay medium, Biotin standard, Cover plastic foil, Replacement holder for 96-well plates, Biotin control 1 and 2.