L(-)-Glutathione (reduced form) 98.0-101.0% (by iodometry), EMPROVE® EXPERT Ph. Eur. for biopharmaceutical production
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:70-18-8
:
Formula:C₁₀H₁₇N₃O₆S
MDL Number:MFCD00065939
:307.33 g/mol
Melting Pt:185…195 °C
Density:160 kg/m³
Storage Temperature:Refrigerator
Specification Test Results
- Assay (iodometric, calc. on dried substance):98.0 - 101.0 %
- Identity (IR-spectrum):passes test
- Appearance:white to almost white, slightly agglomerated, crystalline powder
- Appearance of solution (100 g/l, water):clear and colorless.
- Spec. rotation (α 20/D, 40 g/l, water, calc. on dried substance):-15.5° to -17.5°
- Chloride (Cl):≤ 200 ppm
- Sulfate (SO₄):≤ 300 ppm
- As (Arsenic):≤ 1.5 ppm
- Fe (Iron):≤ 10 ppm
- Fe (Iron) (ICP):≤ 10 ppm
- NH₄ (Ammonium):≤ 200 ppm
- Cu (Copper)*:≤ 25 ppm
- Related substances (CE) (Impurity A (Cysteinylglycine)):≤ 0.5 %
- Related substances (CE) (Impurity B (Cysteine)):≤ 0.5 %
- Related substances (CE) (Impurity C (Glutathione oxidized)):≤ 1.5 %
- Related substances (CE) (Impurity D (Glutamylcysteine)):≤ 1.0 %
- Related substances (CE) (Impurity E):≤ 0.5 %
- Related substances (CE) (Largest unspecified impurity):≤ 0.2 %
- Related substances (CE) (Sum of all impurities):≤ 2.5 %
- Acetic acid (HS-GC):≤ 5000 ppm
- Ethanol (HS-GC):≤ 5000 ppm
- Ethyl acetate (HS-GC):≤ 5000 ppm
- Other residual solvents (ICH Q3C):passes test
- Sulfated ash (600 °C):≤ 0.1 %
- Loss on drying (105 °C; 3 h):≤ 0.5 %
- Corresponds to Ph. Eur.:—