Order Entry
Puerto Rico
ContactUsLinkComponent
 
NAD Synthetase (from E. coli)
Catalog # 102980-454
Supplier:  Adipogen
CAS Number:  
NAD Synthetase (from E. coli)
Catalog # 102980-454
Supplier:  Adipogen
Supplier Number:  AG-40T-0109-C050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Size:
    50 µg
  • Storage Conditions:
    –80 °C
  • Form:
    Liquid
  • Shipping Conditions:
    –80 °C, dry ice
  • Enzyme Name:
    NAD Synthetase
  • Enzyme Activity:
    ~0.3 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1µmol NAD per min.
  • Purity:
    98%
  • Molecular Weight:
    ~30.4 kDa
  • Concentration:
    0.5 mg/ml (Lot specific)
  • Formulation:
    In 50 mM TRIS-HCl, pH 8, containing 75 mM sodium chloride, 5% glycerol and 5 mM DTT.
  • Cat. No.:
    102980-454
  • Supplier no.:
    AG40T0109C050

Specifications

About this item

NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD+, catalyzing the final step of both pathways

  • UniProt link P08164: NAD Synthetase (B. subtilis)

Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It was shown to be induced by heat shock, salt stress, oxidative stress, glucose limitation and oxygen limitation and is involved in biological processes like stress response and sporulation resulting in formation of a cellular spore. Catalytic activity: ATP + deamido-NAD+ + NH3 = AMP + diphosphate + NAD+