Order Entry
Puerto Rico
ContactUsLinkComponent
Human NAPRTase (from E. coli)
Human NAPRTase (from E. coli)
Catalog # 102980-446
Supplier:  Adipogen
CAS Number:  
Human NAPRTase (from E. coli)
Catalog # 102980-446
Supplier:  Adipogen
Supplier Number:  AG-40T-0105-C050
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Storage Conditions:
    –80 °C
  • Form:
    Liquid
  • Shipping Conditions:
    –80 °C, dry ice
  • Enzyme Name:
    NAPRTase
  • Enzyme Activity:
    ~0.27 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1µmol nicotinic acid mononucleotide (NaMN) per min.
  • Purity:
    98%
  • Molecular Weight:
    ~58.0 kDa
  • Concentration:
    0.5 mg/ml (Lot specific)
  • Formulation:
    In 50 mM HEPES/KOH pH 7.5 containing 300 mM potassium chloride and 10 mM DTT.
  • Cat. No.:
    102980-446
  • Supplier no.:
    AG40T0105C050

Specifications

About this item

Human nicotinate phosphoribosyltransferase (NAPRTase) is localized in the cytoplasm and is involved in biological processes such as NAD biosynthetic and metabolic processes, nicotinamide metabolic process, nicotinate nucleotide salvage, response to oxidative stress and water-soluble vitamin metabolic process

  • UniProt link Q6XQN6: NAPRTase (human)

It functions by catalyzing the conversion of nicotinic acid (NA) to NA mononucleotide (NaMN) and is essential for NA to increase cellular NAD levels and prevent oxidative stress of the cells. It is a crucial factor in the NAD+ biosynthesis pathway. Catalytic activity: Beta-nicotinate D-ribonucleotide + diphosphate = nicotinate + 5-phospho-alpha-D-ribose 1-diphosphate.