Order Entry
Puerto Rico
ContactUsLinkComponent
Human NAD Kinase (from E. coli)
Human NAD Kinase (from E. coli)
Catalog # 102980-422
Supplier:  Adipogen
CAS Number:  
Human NAD Kinase (from E. coli)
Catalog # 102980-422
Supplier:  Adipogen
Supplier Number:  AG-40T-0091-C050
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Free
  • Form:
    Liquid
  • Shipping Conditions:
    –20 °C, blue Ice
  • Enzyme Name:
    NAD Kinase
  • Enzyme Activity:
    >1 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1 µmol of NADP per min.
  • Purity:
    95%
  • Molecular Weight:
    ~50 kDa (monomer)
  • Concentration:
    1 mg/ml
  • Formulation:
    Ammonium sulfate suspension
  • Cat. No.:
    102980-422
  • Supplier no.:
    AG40T0091C050

Specifications

About this item

NAD kinase catalyzes the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions

  • UniProt link O95544: NAD kinase (human)

NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.