About this item
Stem cell factor (SCF), also known as cKit ligand (KL), mast cell growth factor (MGF) and steel factor (SLF), is a widely expressed 28-40 kDa type I transmembrane glycoprotein. It promotes the survival, differentiation and mobilization of multiple cell types including myeloid, erythroid, megakaryocytic, lymphoid, germ cell and melanocyte progenitors. SCF is a primary growth and activation factor for mast cells and eosinophils. Noncovalent dimers of transmembrane or soluble SCF interact with the receptor tyrosine kinase SCF R/cKit to trigger receptor dimerization and signaling. SCF assists in the recovery of cardiac function following myocardial infarction by increasing the number of cardiomyocytes and vascular channels.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Gene ID:NP_003985.2
- Protein Synonyms:c-Kit Ligand|Mast Cell Grow Factor|Stem Cell Factor|MGF|Kit Ligand
- Protein/Peptide Name:SCF
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice