About this item
Interleukin-8 (IL-8) was originally discovered as a neutrophil chemotactic and activating factor and is a member of the alpha (CXC) subfamily of chemokines (including also platelet factor 4, GRO, IP-10, etc.). Many cell types, including monocyte/macrophages, T cells, neutrophils, fibroblasts, endothelial cells, keratinocytes, hepatocytes, chondrocytes and various tumor cell lines, produce IL-8 in response to a wide variety of proinflammatory stimuli such as exposure to IL-1, TNF, LPS and viruses. IL-8 has a wide range of other proinflammatory effects. It is a potent chemoattractant for neutrophils and causes degranulation of neutrophil specific granules and azurophilic granules. IL-8 induces expression of the cell adhesion molecules CD11/CD18 and enhances the adherence of neutrophils to endothelial cells and subendothelial matrix proteins. Besides neutrophils, IL-8 is also chemotactic for basophils, T cells and eosinophils. IL-8 has been reported to be a co-mitogen for keratinocytes and was also shown to be an autocrine growth factor for melanoma cells. IL-8 was also reported to be angiogenic both in vivo and in vitro.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein Function:Interleukins
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Biological Activity:Shows the biological function of the IL-8 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain.
- Gene ID:NP_000575.1
- Protein Synonyms:NCF|MDNCF|Interleukin-8|C-X-C Motif Chemokine 8|Emoctakin|TCF|LAI|NAP-1|MONAP|GCP-1|CXCL8
- Protein/Peptide Name:IL-8
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice