About this item
Interleukin-4 (IL-4) is a cytokine produced by type 2 helper T cells, the Th2 cells. These cells tends to make a specific set of lymphokines including IL-4, IL-5, IL-6, IL-10, IL-13, IL-3 and GM-CSF and fail to produce IL-2, IFN-gamma, and lymphotoxin (TNF-beta). In addition, mast cells can produce IL-4. IL-4 exerts numerous effects on various hematopoietic cell types. On B cells, IL-4 promotes immunoglobulin class switching to IgE and IgG1 isotypes and upregulates MHC class II and CD23 expression. IL-4 promotes survival, growth, and differentiation of both T and B lymphocytes, mast cells and endothelial cells. In addition, IL-4 inhibits the production of TNF, IL-1, and IL-6 by macrophages.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein Function:Interleukins
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Gene ID:NP_000580
- Protein Synonyms:B Cell Stimulatory Factor 1|BSF-1|Interleukin-4
- Protein/Peptide Name:IL-4
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice