To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:100 µg
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Gene ID:NP_001954.2
- Protein Synonyms:Pro-Epidermal Growth Factor
- Protein/Peptide Name:EGF
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice
- Cat. No.:102981-550
- Supplier no.:HF210EGFC100
Specifications
About this item
Epidermal growth factor (EGF) is a growth factor and the founding member of the EGF family. All EGF family members are synthesized as type I transmembrane precursor proteins that may contain several EGF domains in the extracellular region. The mature proteins are released from the cell surface by regulated proteolysis. EGF is present in various body fluids, including blood, milk, urine, saliva, seminal fluid, pancreatic juice, cerebrospinal fluid, and amniotic fluid. Four ErbB (HER) family receptor tyrosine kinases including EGFR/ErbB1, ErbB2, ErbB3 and ErbB4, mediate responses to EGF family members. These receptors undergo a complex pattern of ligand induced homo or heterodimerization to transduce EGF family signals. EGF binds to the receptor EGFR stimulating the intrinsic protein-tyrosine kinase activity of the receptor. The tyrosine kinase activity initiates a signal transduction cascade that results in a variety of biochemical changes within the cell, including a rise in intracellular calcium levels, increased glycolysis and protein synthesis, and increases in the expression of certain genes including the gene for EGFR, which lead to DNA synthesis, cell growth, proliferation and differentiation. Other biological activities ascribed to EGF include epithelial development, angiogenesis, inhibition of gastric acid secretion, fibroblast proliferation, and colony formation of epidermal cells in culture. Defects in EGF are the cause of hypomagnesemia type 4 (HOMG4), also known as renal hypomagnesemia normocalciuric. HOMG4 is a disorder characterized by massive renal hypomagnesemia and normal levels of serum calcium and calcium excretion. Clinical features include seizures, mild-to mederate psychomotor retardation, and brisk tendon reflexes.