Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CD279 (from HEK293 Cells)
Human Recombinant CD279 (from HEK293 Cells)
Catalog # 102981-536
Supplier:  Adipogen
CAS Number:  
Human Recombinant CD279 (from HEK293 Cells)
Catalog # 102981-536
Supplier:  Adipogen
Supplier Number:  CHI-HF-211PD1-C100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <5EU/mg
  • Gene ID:
    NP_005009.2
  • Protein Synonyms:
    PD-1|Programmed Cell Death Protein 1|SLEB2
  • Protein/Peptide Name:
    CD279
  • Purity:
    98%
  • Molecular Weight:
    ~60kDa (SDS-PAGE)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-536
  • Supplier no.:
    HF211PD1C100

Specifications

About this item

CD279 (Programmed Cell Death Protein 1; PD-1) is a type I transmembrane protein belonging to the CD28/CTLA-4 family of immunoreceptors that mediate signals for regulating immune responses. Members of the CD28/CTLA-4 family have been shown to either promote T cell activation (CD28 and ICOS) or downregulate T cell activation (CTLA-4 and PD-1). CD279 is expressed on activated T cells, B cells, myeloid cells and on a subset of thymocytes. In vitro, ligation of CD279 inhibits TCR-mediated T cell proliferation and production of IL-1, IL-4, IL-10 and IFN-gamma. In addition, CD279 ligation also inhibits BCR mediated signaling. CD279 deficient mice have a defect in peripheral tolerance and spontaneously develop autoimmune diseases.