Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant CD278 (from CHO cells)
Mouse Recombinant CD278 (from CHO cells)
Catalog # 102981-534
Supplier:  Adipogen
CAS Number:  
Mouse Recombinant CD278 (from CHO cells)
Catalog # 102981-534
Supplier:  Adipogen
Supplier Number:  CHI-MF-110ICOS-C100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Gene ID:
    NP_059508.2
  • Protein Synonyms:
    CRP1|CD28-related Protein-1|AILIM|Inducible T Cell Costimulator|ICOS
  • Protein/Peptide Name:
    CD278
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-534
  • Supplier no.:
    MF110ICOSC100

Specifications

About this item

CD278, also called inducible costimulator (ICOS) or CRP1 (CD28-related protein-1), is a member of the growing CD28 family of immune costimulatory receptors. Other family members are CD28, CTLA-4 and PD-1. ICOS is expressed on most CD45RO+ cells. ICOS expression is upregulated within approximately 24-48 hours of activation on Th primed cells. B7-H2, a member of the B7 family of costimulatory ligands, has been identified as the ICOS ligand. The B7-H2/ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.