Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant CD275 (from CHO cells)
Catalog # 102981-516
Supplier:  Adipogen
CAS Number:  
Mouse Recombinant CD275 (from CHO cells)
Catalog # 102981-516
Supplier:  Adipogen
Supplier Number:  CHI-MF-120B7H2-C100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized human ICOS at 1µg/ml (100µl/well) can bind biotinylated B7-H2 (mouse):Fc (mouse) (rec.) (non-lytic) with a linear range of 0.1-1.0µg/ml. Optimal dilutions should be determined by each laboratory for each application.
  • Gene ID:
    NP_056605.1
  • Protein Synonyms:
    ICOS Ligand|B7-H2|B7RP-1|B7-like Protein Gl50
  • Protein/Peptide Name:
    CD275
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-516
  • Supplier no.:
    MF120B7H2C100

Specifications

About this item

Mouse CD275 (B7-H2) is a member of the growing B7 family of immune costimulatory proteins. CD275 has been identified as the ligand for ICOS, a member of the CD28 family of co-stimulatory receptors. Mouse CD275 is expressed on resting B cells and at low levels on monocytes. The CD275/ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.