Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant CD273 (from CHO cells)
Mouse Recombinant CD273 (from CHO cells)
Catalog # 102981-506
Supplier:  Adipogen
CAS Number:  
Mouse Recombinant CD273 (from CHO cells)
Catalog # 102981-506
Supplier:  Adipogen
Supplier Number:  CHI-MF-110PDL2-C100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Gene ID:
    NP_067371.1
  • Protein Synonyms:
    Programmed Cell Death 1 Ligand 2|PDL2|B7-DC|PDCD1 Ligand 2
  • Protein/Peptide Name:
    CD273
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-506
  • Supplier no.:
    MF110PDL2C100

Specifications

About this item

T cells require a signal induced by the engagement of the T cell receptor and a costimulatory signal(s) through distinct T cell surface molecules for optimal T cell activation and tolerance. CD273 (PD-L2) is one of two ligands for programmed death-1 (PD-1; CD279), a member of the CD28 family of immunoreceptors. The other identified ligand is PD-L1. CD273 is broadly expressed and also up regulated in a variety of tumor cell lines. On previously activated T cells, CD273 interaction with PD-1 inhibits TCR mediated proliferation and cytokine production, suggesting an inhibitory role in regulating immune responses. CD273 has a costimulatory function on resting T cells activated with suboptimal TCR signals.