Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CD273 (from CHO Cells)
Human Recombinant CD273 (from CHO Cells)
Catalog # 102981-502
Supplier:  Adipogen
CAS Number:  
Human Recombinant CD273 (from CHO Cells)
Catalog # 102981-502
Supplier:  Adipogen
Supplier Number:  CHI-HF-210PDL2-C100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Gene ID:
    NP_079515.2
  • Protein Synonyms:
    Programmed Cell Death 1 Ligand 2|PDL2|B7-DC|PDCD1 Ligand 2
  • Protein/Peptide Name:
    CD273
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-502
  • Supplier no.:
    HF210PDL2C100

Specifications

About this item

T cells require a signal induced by the engagement of the T cell receptor and a costimulatory signal(s) through distinct T cell surface molecules for optimal T cell activation and tolerance. CD273 (PD-L2) is one of two ligands for programmed death-1 (PD-1; CD279), a member of the CD28 family of immunoreceptors. The other identified ligand is PD-L1. CD273 is broadly expressed and also up regulated in a variety of tumor cell lines. On previously activated T cells, CD273 interaction with PD-1 inhibits TCR mediated proliferation and cytokine production, suggesting an inhibitory role in regulating immune responses. CD273 has a costimulatory function on resting T cells activated with suboptimal TCR signals.