Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CD137 (from CHO cells)
Human Recombinant CD137 (from CHO cells)
Catalog # 102981-484
Supplier:  Adipogen
CAS Number:  
Human Recombinant CD137 (from CHO cells)
Catalog # 102981-484
Supplier:  Adipogen
Supplier Number:  CHI-HF-210CD137-C100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Biological Activity:
    Measured by its binding ability in a functional ELISA.
  • Gene ID:
    AAH06196.1
  • Protein Synonyms:
    CD137 Antigen|Tumor Necrosis Factor Receptor Superfamily Member 9|T Cell Antigen ILA|TNFRSF9|MGC2172|ILA|CDw137|4-1BB|4-1BB Ligand Receptor T Cell|Antigen 4-1BB Homolog
  • Protein/Peptide Name:
    CD137
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-484
  • Supplier no.:
    HF210CD137C100

Specifications

About this item

Human CD137 (4-1BB) is a costimulatory molecule of the tumor necrosis factor (TNF) receptor superfamily. The glycoprotein 4-1BB is expressed mainly on activated CD4+ and CD8+ T cells and binds to a high-affinity ligand (4-1BBL) expressed on several antigen-presenting cells such as macrophages and activated B cells. Upon ligand binding, 4-1BB is associated with the tumor receptor-associated factors (TRAF), the adaptor protein and mediates downstream signaling events including the activation of NF-kappaB and cytokine production. 4-1BB signaling either by binding to 4-1BBL or by antibody ligation delivers signals for T cell activation and growth as well as monocyte proliferation and B cell survival, and plays a important role in the amplification of T cell-mediated immune responses. In contrast, it can also enhance activation-induced T cell apoptosis when triggered by engagement of the TCR/CD3 complex. In addition, the 4-1BB/4-1BBL costimulatory pathway has been shown to augment secondary CTL responses to several viruses and increase antitumor immunity. 4-1BB is therefore a promising candidate for immunotherapy of human cancer.