Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant CD134 (from CHO cells)

Human Recombinant CD134 (from CHO cells)

50 µg
Catalog # 102981-480
Marketsource
Supplier:   Adipogen
Supplier #:  CHI-HF-210CD134-C050
Human Recombinant CD134 (from CHO cells)
Catalog # 102981-480
Supplier:  Adipogen
Marketsource
CAS Number:  
Human Recombinant CD134 (from CHO cells)

50 µg

CAS Number:  
Price
$443.08
Log in for availability
Each (50µG)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CD134 (OX40) is a T cell activation antigen structurally belonging to a lymphocyte-specific subgroup of the nerve growth factor and tumor necrosis factor receptor superfamily, which also includes the T cell antigen CD27, B cell antigen CD40, FAS antigen and the T cell activation antigen 4-1BB. The human CD134 protein is expressed only on activated CD4+ T blasts, and its ligand has been identified as gp34. Interactions between CD134 and its ligand in vivo are necessary for the differentiation of activated B cells into highly immunoglobulin-producing cells, however not involved in other pathways of antigen-driven differentiation of B cells such as development of memory cells in the germinal centers. In addition, the CD134 and gp34 system directly mediate adhesion of activated T cells to vascular endothelial cells, and contribute to growth stimulation of the virus-infected T cells.

Specifications

Specifications

  • Size:
    50 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Level:
    Low
  • Purity:
    98%
  • Endotoxin Content:
    <0.06EU/µg
  • Source:
    CHO cells
  • Cat. No.:
    102981-480
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Biological Activity:
    Measured by its binding ability in a functional ELISA.
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    NP_003318.1
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Protein Synonyms:
    TXGP1L|ACT35|Tumor Necrosis Factor Receptor Superfamily Member 4|OX40|TNFRSF4
  • Protein/Peptide Name:
    CD134
  • Conjugation:
    Unconjugated
  • Species:
    Human

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.