Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant CD86 (from CHO cells)
Mouse Recombinant CD86 (from CHO cells)
Catalog # 102981-478
Supplier:  Adipogen
CAS Number:  
Mouse Recombinant CD86 (from CHO cells)
Catalog # 102981-478
Supplier:  Adipogen
Supplier Number:  CHI-MF-110CD86-C100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Gene ID:
    NP_062261.3
  • Protein Synonyms:
    B70|ETC-1|B7-2
  • Protein/Peptide Name:
    CD86
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-478
  • Supplier no.:
    MF110CD86C100

Specifications

About this item

CD86 (B7-2) is a 60-100 kDa variably glycosylated protein in the B7 family. B7 family members are transmembrane cell surface molecules that play important roles in immune activation and the maintenance of immune tolerance. B7-2 is highly expressed on activated antigen presenting cells (APC), e.g. B cells, dendritic cells and monocytes as well as on vascular endothelial cells. B7-2 and the closely related CD80 (B7-1) exhibit overlapping but distinct functional properties. Their binding to CD28, which is constitutively expressed on T cells, enhances T cell receptor signaling and also provides TCR-independent costimulation. B7-1 and B7-2 additionally bind the CD28-related protein CTLA-4, which is up-regulated and recruited to the immunological synapse (IS) at the onset of T cell activation. CTLA-4 ligation inhibits the T cell response and supports regulatory T cell function. B7-2 is expressed earlier than B7-1 following APC activation and both proteins bind with higher affinity to CTLA-4 than to CD28. B7-2 promotes the stabilization of CD28 in the IS, while B7-1 is primarily responsible for promoting CTLA-4 recruitment and accumulation in the IS. The relative participation of B7-1 and B7-2 in T cell costimulation can also alter the Th1/Th2 bias of the immune response. Both B7-1 and B7-2 serve as cellular receptors for B species adenoviruses.