Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CD40 (from CHO Cells)
Human Recombinant CD40 (from CHO Cells)
Catalog # 102981-464
Supplier:  Adipogen
CAS Number:  
Human Recombinant CD40 (from CHO Cells)
Catalog # 102981-464
Supplier:  Adipogen
Supplier Number:  CHI-HF-210CD40-C100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Biological Activity:
    Measured by its binding ability in a functional ELISA assay.
  • Gene ID:
    AAH12419.1
  • Protein Synonyms:
    CD40L Receptor|Tumor Necrosis Factor Receptor Superfamily Member 5|TNFRSF5
  • Protein/Peptide Name:
    CD40
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102981-464
  • Supplier no.:
    HF210CD40C100

Specifications

About this item

CD40 is a member of the TNF receptor superfamily which are single transmembrane-spanning glycoproteins and plays an essential role in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. CD40 contains 4 cysteine-rich repeats in the extracellular domain and is expressed in B cells, dendritic cells, macrophages, endothelial cells and several tumor cell lines. The cognate interaction between CD40 and CD40 ligand (CD154) on T cells activates NF-kappaB, Jun N-terminal kinase and Janus kinase signal transducers and activators of transcription pathways. Several different TRAF proteins (adapter proteins) have been identified to serve as mediators of the signal transduction. In addition, CD40/CD40L interaction is found to be necessary for amyloid-beta-induced microglial activation and thus is thought to be an early event in Alzheimer disease pathogenesis. Defects in CD40 result in hyper-IgM immunodeficiency type 3 (HIGM3), an autosomal recessive disorder characterized by the inability of B cells to undergo isotype switching, as well as an inability to mount an antibody-specific immune response and a lack of germinal center formation.