About this item
CCL22, also named stimulated T cell chemotactic protein (STCP1), is a CC chemokine initially isolated from clones of monocyte-derived macrophages and binds to the receptor CCR4. Human CCL22 is highly expressed in macrophage and in monocyte-derived dendritic cells and thymus. It is also found in lymph node, appendix, activated monocytes, resting and activated macrophages. May play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. CCL22 has been shown to induce chemotaxis or Ca2+ mobilization in dendritic cells, IL-2 activated NK cells and activated T lymphocytes.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Chemokine
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Gene ID:AAH27952.1
- Protein Synonyms:MDC|Macrophage-derived Chemokine|C-C Motif Chemokine 22|CC Chemokine STCP-1|Stimulated T Cell Chemotactic Protein 1|Small-inducible Cytokine A22
- Protein/Peptide Name:CCL22
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice