Order Entry
Puerto Rico
ContactUsLinkComponent
Human CD137L (from CHO cells)
Human CD137L (from CHO cells)
Catalog # 102513-228
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Human CD137L (from CHO cells)
Catalog # 102513-228
Supplier:  Adipogen
Supplier Number:  ANC-503-820
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    25 µg
  • Storage Conditions:
    +4 °C
  • Protein Synonyms:
    4-1BBL|Tumor Necrosis Factor Ligand Superfamily Member 9|TNFSF9
  • Protein/Peptide Name:
    CD137L
  • Formulation:
    50 mM Sodium Phosphate pH 7.5, 100 mM Potassium Chloride, 150mM NaCl
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102513-228
  • Supplier no.:
    ANC503820

Specifications

About this item

Human CD137L (4-1BB ligand) is a type II transmembrane protein constitutively expressed by monocytes, B cells and neuroblastoma cells. Binding of CD137 (4-1BB) to CD137L induces monocyte activation.