Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant POP (from <i>E. coli</i>)

Human Recombinant POP (from E. coli)

Marketsource
Supplier:   Adipogen
Human Recombinant POP (from <i>E. coli</i>)
Supplier:  Adipogen
Marketsource
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The inflammasome is a multiprotein complex that mediates the activation of caspase-1, which promotes, amongst others, the secretion of the proinflammatory cytokines interleukin (IL)-1beta and IL-18. Members of the Nod-like receptor family (NLRs), including NLRP3/NALP3, are critical components of the inflammasome that link danger-signals to caspase-1 activation. Defects in NLRP3 are the cause of familial cold autoinflammatory syndrome type 1 (FCAS1), Muckle-Wells syndrome (MWS) and of chronic infantile neurologic cutaneous and articular syndrome (CINCA). Pyrin domain-containing protein 1 (POP1) interacts with the protein Asc (Pycard) and modulates the interaction of Asc to NLR proteins.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 102980-212
  • 102980-214
  • Protein/Peptide Name:
  • POP
  • POP
  • Protein Synonyms:
  • PYC1|Pyrin Domain-containing Protein 1|Pyrin-only Protein 1|PAAD-only Protein 1
  • PYC1|Pyrin Domain-containing Protein 1|Pyrin-only Protein 1|PAAD-only Protein 1
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • Q8WXC3
  • Q8WXC3
  • Purity:
  • 95%
  • 95%
  • Concentration:
  • 0.1mg/ml after reconstitution.
  • 0.1mg/ml after reconstitution.
  • Formulation:
  • Lyophilized. Contains PBS.
  • Lyophilized. Contains PBS.
  • Molecular Weight:
  • ~38 kDa
  • ~38 kDa
  • Storage Conditions:
  • –20 °C
  • –20 °C
  • Shipping Temperature:
  • -20 °C, Blue Ice
  • -20 °C, Blue Ice

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 10 µg
    Log in for availability
    Log in for availability
    Each (10µG)
    Each (10µG) - $207.69
    $207.69
    Each (10µG)
       $207.69

    Supplier Number: AG-40B-0123-C010
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102980-212
    Protein/Peptide Name
    POP
    Protein Synonyms
    PYC1|Pyrin Domain-containing Protein 1|Pyrin-only Protein 1|PAAD-only Protein 1
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q8WXC3
    Purity
    95%
    Concentration
    0.1mg/ml after reconstitution.
    Formulation
    Lyophilized. Contains PBS.
    Molecular Weight
    ~38 kDa
    Storage Conditions
    –20 °C
    Shipping Temperature
    -20 °C, Blue Ice
    Marketsource
  • 3 x 10 µg
    Log in for availability
    Log in for availability
    Pack of 3 (10µµG)
    Pack of 3 (10µµG) - $415.38
    $415.38
    Pack of 3 (10µµG)
       $415.38

    Supplier Number: AG-40B-0123-3010
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102980-214
    Protein/Peptide Name
    POP
    Protein Synonyms
    PYC1|Pyrin Domain-containing Protein 1|Pyrin-only Protein 1|PAAD-only Protein 1
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q8WXC3
    Purity
    95%
    Concentration
    0.1mg/ml after reconstitution.
    Formulation
    Lyophilized. Contains PBS.
    Molecular Weight
    ~38 kDa
    Storage Conditions
    –20 °C
    Shipping Temperature
    -20 °C, Blue Ice
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.