Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant IL-36alpha (from E. coli)
Mouse Recombinant IL-36alpha (from E. coli)
Catalog # 102980-144
Supplier:  Adipogen
CAS Number:  
Mouse Recombinant IL-36alpha (from E. coli)
Catalog # 102980-144
Supplier:  Adipogen
Supplier Number:  AG-40B-0098-C010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Interleukins
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    10 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.01EU/µg
  • Biological Activity:
    Induces secretion of IL-6 in BMDCs at a concentration of 100ng - 1µg/ml.
  • Gene ID:
    Q9JLA2
  • Amino Acid Number:
    8 to 160
  • Protein Synonyms:
    Interleukin-1 Family Member 6|Interleukin-36alpha|IL-1H1|Interleukin-1epsilon|FIL1epsilon|IL-1F6|Interleukin-1 Homolog 1
  • Protein/Peptide Name:
    IL-36alpha
  • Purity:
    95%
  • Molecular Weight:
    ~17kDa
  • Endotoxin Level:
    Low
  • Concentration:
    0.1mg/ml after reconstitution.
  • Formulation:
    Lyophilized. Contains PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102980-144
  • Supplier no.:
    AG40B0098C010

Specifications

About this item

IL-36alpha (IL-1F6), IL-36beta (IL-1F8) and IL-36gamma (IL-1F9) bind to IL-36R (IL-1Rrp2) and IL-1RAcP, activating similar intracellular signals as IL-1 and are inhibited by IL-36Ra. The expression of IL-36 cytokines has been shown to occur at different sites including the lung and skin and can be derived from diverse cell types including keratinocytes, bronchial epithelium as well as macrophages, monocytes and different T cell subsets. IL-36 family members induce the production of proinflammatory cytokines, including IL-12, IL-1beta, IL-6, TNF-alpha and IL-23 in BMDC and CD4 T cells. Skin and dendritic cells are targets of the IL-36 interleukins leading to a Th1 response. These cytokines may represent potential targets for immune-mediated inflammatory conditions or, alternatively, could be used as adjuvants in vaccination. Recently a novel role for IL-36alpha in the pathogenesis of intestinal inflammation has been reported.