Tumor necrosis factor (TNF, cachexin or cachectin and formerly known as tumor necrosis factor-alpha) is a cytokine involved in systemic inflammation and is a member of a group of cytokines that stimulate the acute phase reaction. MultimericTNF-alpha™ is a high activity construct in which two trimeric TNF-alpha ligands are artificially linked via the collagen domain of ACRP30. The receptor TNF-R1 is activated by both the membrane-bound and soluble trimeric forms of TNF-alpha, whereas the receptor TNF-R2 only responds to the membrane-bound form of TNF-alpha (TNF-alpha (mouse) (multimeric) (rec.) (Prod. No. AG-40B-0021). Since the MultimericTNF-alpha™ mimics the membrane-bound form (forms oligomers higher than trimer), it is the only TNF-alpha protein that can activate the TNF-R2. For TNF-R1 activation, either "normal" TNF-alpha or MultimericTNF-alpha™ can be used.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:<0.02EU/µg
- Biological Activity:Activates human and mouse TNF-R1 and TNF-R2. Induces cell death at a concentration range of 0.01-10ng/ml (ED50= 0.05ng/ml or 2 x 10E7 Units/mg; WEHI 164 cells).
- Gene ID:P06804
- Protein Synonyms:ACRP30headless:TNF-alpha|ACRP30headless:Tumor Necrosis Factor-alpha|ACRP30headless:TNFSF2|MultimericTNF-alpha™
- Protein/Peptide Name:TNF-alpha
- Purity:95%
- Molecular Weight:~34kDa (SDS-PAGE)
- Endotoxin Level:Low
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Shipping Temperature:-20 °C, Blue Ice