- Size:100 µg
- Protein/Peptide Name:RED fluorogenic Substrate
- Purity:>90% (HPLC)
- Molecular Weight:1910.7
- Sequence:TQ3-GABA-Pro-Cha-Abu-Smc-His-Ala-Dab(6'-TAMRA)-Ala-Lys-NH2
- Cat. No.:76002-134
Long emission wavelength reduces background
TQ3-GABA-Pro-Cha-Abu-Smc-His-Ala-Dab(6'-TAMRA)-Ala-Lys-NH2
[TQ3=quencher; GABA=4-aminobutyric acid; Cha=L-cyclohexylalanine; Abu=2-aminobutyric acid; Smc=S-methyl-L-cysteine; Dab=2,4-diaminobutyric acid; 6’-TAMRA=6’-tetramethylrhodamine]
Highly-quenched, ultra-bright fluorogenic substrate for most MMPs. 6’-TAMRA fluorescence is thoroughly quenched by the TQ3 group until cleavage by MMPs separates the two moieties. The OMNIMMP® RED substrate offers key advantages over other MMP substrates.
Highly-quenched, ultra-bright fluorogenic substrate for most MMPs. 6’-TAMRA fluorescence is thoroughly quenched by the TQ3 group until cleavage by MMPs separates the two moieties. The OMNIMMP® RED substrate offers key advantages over other MMP substrates.
TQ3-GABA-Pro-Cha-Abu-Smc-His-Ala-Dab(6'-TAMRA)-Ala-Lys-NH2
[TQ3=quencher; GABA=4-aminobutyric acid; Cha=L-cyclohexylalanine; Abu=2-aminobutyric acid; Smc=S-methyl-L-cysteine; Dab=2,4-diaminobutyric acid; 6’-TAMRA=6’-tetramethylrhodamine]
Highly-quenched, ultra-bright fluorogenic substrate for most MMPs. 6’-TAMRA fluorescence is thoroughly quenched by the TQ3 group until cleavage by MMPs separates the two moieties. The OMNIMMP® RED substrate offers key advantages over other MMP substrates.