Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AZD-8055
AZD-8055 ≥98% (by HPLC)
AZD-8055 ≥98% (by HPLC)
AZD-8055 ≥98% (by HPLC)
Supplier:
Enzo Life Sciences
Certificates
Expand image
AZD-8055 ≥98% (by HPLC)
Supplier:
Enzo Life Sciences
About this item
CAS Number:
1009298-09-2
:
Formula:
C₂₅H₃₁N₅O₄
MDL Number:
MFCD16660191
:
465.5 g/mol