Glycogen phosphorylase inhibitor ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:648926-15-2
:
Formula:C₁₇H₁₅ClF₂N₄O₄
:412.8 g/mol
Frequently Bought Together
neodisher® Detergents, Miele
Supplier: Miele
Reusable Filter Animal Cage Covers, Tecniplast®
Supplier: Tecniplast Usa
PVC Film Wrap, T FRANK MCCALLS INC
Supplier: TFrank McCalls
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
SHOE COVER WATER RESIST BLUE 12-15 CS150
Supplier: ULINE MS
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Supplier: Thermo Fisher Scientific
COVER SHOE STD SZ 6-11 CS150
Supplier: ULINE MS
CytoFLEX Sheath Fluid, Beckman Coulter
Supplier: Beckman Coulter
MARKER 6.14 MAURICE CIEF PI 210UL
Supplier: PROTEIN SIMPLE MS
PADS LIGHT DUTY CLEANING SYSTEM 10IN
Supplier: ULINE MS
Amersham™ ECL™ Western Blotting Detection Reagent, Cytivia
Supplier: Cytiva
RINSE PORT COVER
Supplier: SHIMADZU SCIENTIFIC INSTRUMENTS MS
UNDERPAD HIGH-ABSORBANCE 23X36IN CS72
Supplier: COVETRUS NORTH AMERICA MS
EDVOTEK Dual Gel Electrophoresis Apparatus
Supplier: Edvotek
SUTURE ABSORBABLE 3-0 / C-14 / 30IN
Supplier: COVETRUS NORTH AMERICA MS