Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
IQ-1
IQ-1 ≥98% (by TLC)
IQ-1 ≥98% (by TLC)
IQ-1 ≥98% (by TLC)
Supplier:
Enzo Life Sciences
Certificates
Expand image
IQ-1 ≥98% (by TLC)
Supplier:
Enzo Life Sciences
About this item
CAS Number:
331001-62-8
:
Formula:
C₂₁H₂₂N₄O₂
MDL Number:
MFCD00354369
:
362.43 g/mol