Order Entry
Puerto Rico
ContactUsLinkComponent
R-(-)-Glucoraphanin potassium salt ≥99% (by HPLC)
R-(-)-Glucoraphanin potassium salt ≥99% (by HPLC)
Catalog # 76001-998
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
R-(-)-Glucoraphanin potassium salt ≥99% (by HPLC)
Catalog # 76001-998
Supplier:  Enzo Life Sciences
Supplier Number:  350-181-M010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    10 mg
  • Cat. No.:
    76001-998

Specifications

About this item

 :
 :475.58 g/mol (anhy)
CAS Number:21414-41-5
MDL Number:MFCD08460269