Recombinant Dengue NS1 ST2
Catalog # 76003-312
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Size:100 µg
- Protein Synonyms:non-structural protein|Thailand/16681/84|viral|DENV1-NS1
- Protein/Peptide Name:Dengue NS1 ST2
- Purity:>95% (SDS-PAGE)
- Formulation:Liquid. In DPBS pH 7.4.
- Purification:Purified protein.
- Cat. No.:76003-312
Specifications
About this item
Dengue virus NS1 protein serotype 2 presented in its native folded state with all post-translational modifications to deliver optimal antigenicity.
Produced in HEK 293 cells. Dengue Viral NS1 Serotype 2 (DENV2-NS1) from strain Thailand/16681/84.