Corynebacterium diphtheriae Diphtheria toxin (Corynebacterium diphtheriae alpha)
Catalog # 76002-114
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Species:Corynebacterium diphtheriae
- Size:1 mg
- Protein/Peptide Name:Diphtheria toxin (Corynebacterium diphtheriae alpha)
- Concentration:1µg/µl
- Formulation:Lyophilized
- Cat. No.:76002-114
Specifications
About this item
From Corynebacterium diphtheriae.