Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TNF-alpha
- Clonality:Monoclonal
- Clone:J1D9
- Conjugation:Biotin
- Flow Cytometry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:liquid
- Antigen Synonyms:TNFSF2|Cachectin|Tumor Necrosis Factor Ligand Superfamily Member 2
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Recombinant human TNF alpha.
- Cat. No.:102513-180
- Supplier no.:ANC398030
Specifications
About this item
TNF (Tumor necrosis factor; cachexin; cachectin and formerly known as tumor necrosis factor-alpha) is a cytokine involved in systemic inflammation and is a member of a group of cytokines that stimulate the acute phase reaction.
Type: Primary
Antigen: TNF-alpha
Clonality: Monoclonal
Clone: J1D9
Conjugation: Biotin
Epitope:
Host:
Isotype: Mouse IgG1
Reactivity: Human