To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD127
- Clonality:Monoclonal
- Clone:ANC8F2
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:120 tests
- Form:liquid
- Antigen Synonyms:IL-7Ralpha|IL7RA|Interleukin-7 Receptor Subunit alpha
- Storage Temperature:+4°C
- Shipping Temperature:-20° C
- Immunogen:Recombinant human CD127.
- Cat. No.:102513-010
- Supplier no.:ANC348050
Specifications
About this item
CD127 (IL-7Ra) is the specific receptor component for the cytokine interleukin-7 (IL-7). It is found on a wide variety of hematopoietic cell types including B cell precursors and the majority of T cells. Its expression levels are decreased on T cells following activation. CD127 can dimerize with CD132 (IL-2Rgamma) to form a high affinity IL-7 receptor. CD127 engagement is necessary for T cell development in humans.
Type: Primary
Antigen: CD127
Clonality: Monoclonal
Clone: ANC8F2
Conjugation: R-PE
Epitope:
Host:
Isotype: Mouse IgG1kappa
Reactivity: Human